Infusion filters set, Intrapur® Inline
Lieferant: B. Braun
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
4099842N
BRAU4099842NEA 705 EUR InStock
BRAU4099842N
Infusion filters set, Intrapur® Inline
Infusionen
Ready-to-use set with infusion filter.
  • PVC free
  • Retention of bacteria and particles
  • High throughput
Specifically designed for use during chemotherapy.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge