Safety needles, Sterican®
Lieferant: B. Braun
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
4670005S-01 4670035S-01 4670012S-01 4670016S-01 4670025S-01 4670002S-01 4670008S-01 4670045S-01 4670050S-01 4670052S-01 4670055S-01
BRAU4670005S-01EA 95.7 EUR InStock
BRAU4670005S-01 BRAU4670035S-01 BRAU4670012S-01 BRAU4670016S-01 BRAU4670025S-01 BRAU4670002S-01 BRAU4670008S-01 BRAU4670045S-01 BRAU4670050S-01 BRAU4670052S-01 BRAU4670055S-01
Safety needles, Sterican®
Injektionsnadeln
This single-use needle with safety protection is compatible with all standard Luer lock and Luer slip syringes.
  • Made of stainless chromium-nickel steel
  • Needle hub made of PP
The safety feature of Sterican® safety needle is in compliance with the EU Directive and provides an intuitive, one-handed activation. It securely and irreversibly locks the needle within the integrated shield for a safe disposal.
Zertifizierungen: In compliance with EU Directive 2010/32/EU. Colour coding according to ISO 6009.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge