Thermal transfer printer labels, type B-423
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
062374 030127 030131 030134 030137 018651 030232 030245 030508 030104 030118 030239 030148 030221 132414 030510 030218 030357 030124 030105 030235 030212 030151 030117 018658 030106
BRDY062374EA 188.41 EUR InStock
BRDY062374 BRDY030127 BRDY030131 BRDY030134 BRDY030137 BRDY018651 BRDY030232 BRDY030245 BRDY030508 BRDY030104 BRDY030118 BRDY030239 BRDY030148 BRDY030221 BRDY132414 BRDY030510 BRDY030218 BRDY030357 BRDY030124 BRDY030105 BRDY030235 BRDY030212 BRDY030151 BRDY030117 BRDY018658 BRDY030106
Thermal transfer printer labels, type B-423
Etiketten
Polyester labels for thermal transfer printers.
  • Die-cut labels are supplied on a roll
  • Contains some adhesive backed wire marking materials, and also labels that are pre-printed with headers or templates
  • Supplied with the ribbon separate, not in the same enclosure as the tape/sleeves
This material is UL, CSA, and AGA approved with the respective ribbon. See the recommended ribbon series for the appropriate ribbon to use.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge