Portable/wall group lockout box, Prinzing
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
046134
BRDY046134EA 153.79 EUR InStock
BRDY046134
Portable/wall group lockout box, Prinzing
Sicherheitsverriegelungen
This box accommodates 8 to 10 locks (depending on lock size).
  • Made of yellow powder-coated metal (mild sheet steel)
  • Can be used alone or in combination with Industrial Strength lockout station
Bestellinformation: Contents not included.
Order Now
PRODUKT
TECHNISCHE DATEN
EGÄNZENDE PRODUKTE
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge