Labels for BMP™ 71 printer, type B-483, ToughBond
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
115114
BRDY115114EA 32.85 EUR InStock
BRDY115114
Labels for BMP™ 71 printer, type B-483, ToughBond
Etiketten
These polyester labels are ideal for smooth, rough or highly textured/low surface energy surfaces.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge