Preprinted Tags "Energy Source, Electrical"
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
138399 138401 138402 138403 138404 138447 138448 138449 138450 138451 138829 138830 138831 138832 138833 138834 138835 138836 138837 138838
BRDY138399EA 57.91 EUR InStock
BRDY138399 BRDY138401 BRDY138402 BRDY138403 BRDY138404 BRDY138447 BRDY138448 BRDY138449 BRDY138450 BRDY138451 BRDY138829 BRDY138830 BRDY138831 BRDY138832 BRDY138833 BRDY138834 BRDY138835 BRDY138836 BRDY138837 BRDY138838
Preprinted Tags "Energy Source, Electrical"
Etiketten
These electrical energy source tags are constructed of rigid PP with round corners.
  • Permanent acrylic adhesive
  • Helps the lockout process quicker and easier for higher safety
Used to mark your facility's points of control of control for energy sources.
Colour: Red on white
Zertifizierungen: ATEX
Verpackung: 25 tags per pack.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge