Labels for BMP™ 51/53 printer, type B-422
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
143239
BRDY143239EA 44.14 EUR InStock
BRDY143239
Labels for BMP™ 51/53 printer, type B-422
Etiketten
These labels are designed to withstand solvent exposure. They can be used for component identification, punch block and patch panel labels, asset and inventory tracking.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge