Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Labels for BBP33 printers, type B-724

Lieferant: Brady

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

133872 140723 140724 152847 152842 152913 152909 152910 152911 152908 152912 152840
BRDY133872EA 428.76 EUR InStock
BRDY133872 BRDY140723 BRDY140724 BRDY152847 BRDY152842 BRDY152913 BRDY152909 BRDY152910 BRDY152911 BRDY152908 BRDY152912 BRDY152840
Labels for BBP33 printers, type B-724
Etiketten
Amber high temperature polyimide with an ultra-durable adhesive withstands the extreme wash protocol and cleaning chemicals used in printed circuit board manufacturing.

  • Resists high temperatures up to 300 °C
  • Survives on the top or bottom side of printed circuit boards through the solder process
  • Service temperature: –70 to 270 °C

Withstands multiple cycles of harsh condition washes for printed circuit boards and electronic component preprocess labelling.

Zertifizierungen: Fulfills requirements for MIL-STD-202G method 215K and SAE AS81531 marking of electrical insulating material when used with R4300 series ribbons.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge