Labels for BBP33 Printers, B-8591
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
152941 152931 152946 152936 152937 152932 152942 152947
BRDY152941EA 342.41 EUR InStock
BRDY152941 BRDY152931 BRDY152946 BRDY152936 BRDY152937 BRDY152932 BRDY152942 BRDY152947
Labels for BBP33 Printers, B-8591
Etiketten
B-8591 outdoor, harsh-environment labels resist UV, abrasions, high temperatures, fluids and washdowns, and are an ideal print-on-demand thermal transfer label solution.
  • No overlaminate means durability is drastically improved without adding an overlaminate over the print
  • Abrasion-, chemical-, corrosion-, fade-, humidity-, UV-, weather-resistant
  • Quick and easy application with pressure-sensitive adhesive
  • Meets regulatory specifications for halogen-free applications
  • A ribbon is required to print this material
12+ year outdoor durability reduces identification failures and the costs associated with losing this information.
Used for barcode labeling, component and equipment labelling, data and telecommunications labelling, facility identification, inventory and inspection labelling.
Zertifizierungen: Compliant with Directive 2011/65/EU and 2015/863.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge