ToughStripe® Max B-543 Floor Marking Striped Tape
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
170075
BRDY170075EA 204.44 EUR InStock
BRDY170075
ToughStripe® Max B-543 Floor Marking Striped Tape
Klebebänder
ToughStripe® Max is designed to withstand wear and tear in high-traffic areas where sliding or dragging forces often occur, while maintaining the adhesive capabilities you've come to expect from ToughStripe.
  • Provides greater thickness for resistance to cuts and scrapes in high-traffic areas
  • Durability in forklift and industrial vehicle traffic areas
  • Permanent acrylic adhesive, gloss finish
  • Aggressive adhesive performance when you need it, easy removal when you don't
  • Tapered edges to minimise 'pick points' and traffic bumps
  • Easy installation with minimal tools
  • Minimum application temperature: 10 °C
  • Service temperature: −18 to +60 °C
Brady B-543 is used to mark aisles, passageways, storage locations, point out safety equipment and exits, and enforce protective equipment requirements. It is designed for high-traffic environments where durability is highly valued.
Application: Floor marking
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge