Printer Ribbons, M710 R6400 Series
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
173189
BRDY173189EA 76.22 EUR InStock
BRDY173189
Printer Ribbons, M710 R6400 Series
Farbbänder
One-color ribbon for printing a single color onto a label.
  • R6400 series resin formulation offers superior chemical and abrasion resistance when printed on the appropriate Brady® label material
  • Recommended for use with materials designed to resist chemicals, such as B-492 frozen surface polyester and B-481 chemical resistant polyester
  • Self-contained M710 series ribbon cartridges insert straight into the printer with ease
M710 Series snap-in print ribbons enable 5 second ribbon supply changes. No calibrating, no threading, no waste, no hassles, and the ink always faces the right direction.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge