Printer Ribbons, M710 R6200 Series
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
173190
BRDY173190EA 76.22 EUR InStock
BRDY173190
Printer Ribbons, M710 R6200 Series
Farbbänder
One-color ribbon for printing a single color onto a label.
  • R6200 series ribbons offer superior scratch, smear, heat, and chemical resistance when printed on the appropriate Brady® label material
  • Recommended for printing on glossy, smooth, or semi-matte label stock, such as B-498 repositionable vinyl cloth and B-486 metallized polyester
  • Self-contained M710 series ribbon cartridges insert straight into the printer with ease
M710 Series snap-in print ribbons enable 5 second ribbon supply changes. No calibrating, no threading, no waste, no hassles, and the ink always faces the right direction.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge