Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Aggressive Adhesive Multi-Purpose Polyester Labels B-422 for M610, M611, BMP61, M710 (with Media Adapter), BMP71 (with Media Adapter)

Lieferant: Brady

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

173961 174331 174327 174206 174266 174298 174196 174304 174227 174291 173967 174333 174239 174359 174255 173964 174329 174306 174344 173969 174219 173758 174316 173831
BRDY173961EA 85.92 EUR InStock
BRDY173961 BRDY174331 BRDY174327 BRDY174206 BRDY174266 BRDY174298 BRDY174196 BRDY174304 BRDY174227 BRDY174291 BRDY173967 BRDY174333 BRDY174239 BRDY174359 BRDY174255 BRDY173964 BRDY174329 BRDY174306 BRDY174344 BRDY173969 BRDY174219 BRDY173758 BRDY174316 BRDY173831
Aggressive Adhesive Multi-Purpose Polyester Labels B-422 for M610, M611, BMP61, M710 (with Media Adapter), BMP71 (with Media Adapter)
Etiketten
Thermal transfer printable, glossy permanent polyester (B-422) labels are ideal for use in all commercial and industrial applications.

  • Aggressive adhesive good for powder-coated and textured flat surfaces, including low surface energy plastics
  • Designed to withstand exposure to numerous solvents
  • Service temperature of −40 to 100 °C (−40 to 212 °F)
  • Gloss finish

Ideal for barcode labelling, component and equipment labelling, data and telecommunications labelling, electrical labelling, general identification, inventory and inspection labelling applications.

Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R6010 for the M610/BMP61/BradyPrinter M611/M611 label printers or M7-R6000 for the M710/BMP71 label printers. This label is no longer compatible with TLS series printers.

Zertifizierungen: UL recognised. CSA accepted.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge