All-Weather Vinyl Labels for M610, M611, BMP61 Printers
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
174158 174119 174136 174123 174140 174126 174159 174141 174122 174137 174163 174138 174121 174127 174157 174160 174120 174132 174156 174139 174162 174161 174124 174134 174135 174131 174133 174125 174142
BRDY174158EA 66.65 EUR InStock
BRDY174158 BRDY174119 BRDY174136 BRDY174123 BRDY174140 BRDY174126 BRDY174159 BRDY174141 BRDY174122 BRDY174137 BRDY174163 BRDY174138 BRDY174121 BRDY174127 BRDY174157 BRDY174160 BRDY174120 BRDY174132 BRDY174156 BRDY174139 BRDY174162 BRDY174161 BRDY174124 BRDY174134 BRDY174135 BRDY174131 BRDY174133 BRDY174125 BRDY174142
All-Weather Vinyl Labels for M610, M611, BMP61 Printers
Etiketten
This durable, low-shrink vinyl with ultra-aggressive adhesive is the perfect solution for identification on rough surfaces and in harsh environment areas.
  • Permanent adhesive sticks to surfaces that are smooth, rough, or powder coated
  • Will also stick to irregular surfaces or surfaces with low surface energy
  • Clean gloss finish is resistant to chemicals, grease, oil, and cleaning agents, ensuring identification stays legible
  • This material has an average outdoor durability of 8 to 10 years
  • Gloss finish
Vinyl tape is ideal for use in pipe marking, warehouse and safety identification, arc flash and visual workplace labels in both indoor and outdoor use. Labels are ideal for component and equipment labelling, electrical labelling, facility identification, floor marking, laboratory labelling, lean and 5S labelling, pipe and valve marking, and warehouse marking.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part numbers M61-R10010 or M61-R10010-WT.
Zertifizierungen: BS5609:1986 Section 2 and section 3.
Verpackung: One tape per box.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge