Polypropylene Flag Labels for M710, M610 and M611
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
174383 174381
BRDY174383EA 62.63 EUR InStock
BRDY174383 BRDY174381
Polypropylene Flag Labels for M710, M610 and M611
Etiketten
Flags for the M610, M611, and M710 Portable Printers.
  • No visible affect to label when exposed to most common chemicals, oils, or water
  • Service temperature of −70 to 80 °C
Zertifizierungen: UL recognised. CSA accepted. Halogen-free (DIN VDE 0472/part 815).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge