PermaSleeve® Low Smoke Zero Halogen Heat-Shrink Labels for M710 and BMP71
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
174657 174646 174619 174636 174659 174621 174658 174569 174570 174620 174567 174548 174549 174586 174587 174451 174568 174430 174429 174635 174648 174638 174589 174483 174588 174637 174618 174506 174550 174551 174482 174649 174656 174452 174647 174467
BRDY174657EA 194.95 EUR InStock
BRDY174657 BRDY174646 BRDY174619 BRDY174636 BRDY174659 BRDY174621 BRDY174658 BRDY174569 BRDY174570 BRDY174620 BRDY174567 BRDY174548 BRDY174549 BRDY174586 BRDY174587 BRDY174451 BRDY174568 BRDY174430 BRDY174429 BRDY174635 BRDY174648 BRDY174638 BRDY174589 BRDY174483 BRDY174588 BRDY174637 BRDY174618 BRDY174506 BRDY174550 BRDY174551 BRDY174482 BRDY174649 BRDY174656 BRDY174452 BRDY174647 BRDY174467
PermaSleeve® Low Smoke Zero Halogen Heat-Shrink Labels for M710 and BMP71
Etiketten
The B-7641 heat shrink sleeve from Brady® is a low smoke, halogen-free identification material used for wire and cable identification.
  • Made of halogen-free, flexible flame-retardant, heat-shrinkable polyolefin tubing, offering fire safety properties and minimal smoke emission
  • Compatible with the most stringent requirements in the market today to enhance fire safety needs
  • 2:1 heat-shrink ratio
  • Matte finish
  • Use with thermal transfer printers and Brady's R6000 series halogen-free ribbon
Suitable for use in applications that require low smoke, halogen-free, flame retardant markers. Applications include data and telecommunications labelling and wire and cable labelling.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction and better performance and reliability. A ribbon is required to print this material. Use part number M7-R6000.
Zertifizierungen: BS-6853 (1999) Vehicle Category 1A; EN-45545-2, requirement 24, Vehicle Category HL3. Zero Halogen.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge