Rechargeable NiMH Battery for M510/M610/M710 Printers
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
175674
BRDY175674EA 97.71 EUR InStock
BRDY175674
Rechargeable NiMH Battery for M510/M610/M710 Printers
Batterien
Work longer with this NiMH M510, M610 and M710 battery pack. It enables cordless printer operation so you can print labels where (and when) you need them.
  • Removable, rechargeable Nickel-metal hydride battery pack
  • Battery can be charged only while installed in an M510, M610 or M710 printer
  • Prints up to 1,700 labels per charge
Work longer with this NiMH M510, M610 and M710 Battery Pack. It enables cordless printer operation so you can print labels where (and when) you need them. Removable, rechargeable Nickel-metal Hydride Battery Pack. Battery can be charged only while installed in an M510, M610 or M710 printer. Prints up to 1,700 labels per charge.
Bestellinformation: Order reference M-NIMH-BATTERY.
Lieferumfang: NiMH battery pack.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge