Packaging Paper Labels, Picto "This side up"
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
237823
BRDY237823EA 131.14 EUR InStock
BRDY237823
Packaging Paper Labels, Picto "This side up"
Etiketten
Paper labels (B-7610) for facility identification and packaging.
  • White matte paper with black arrows
  • Rectangular
  • Service temperature from -50 to +50° C
1000 labels per pack
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge