Magicard™ Monochrome Black Film + Overcoat for Magicard™ 300
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
321974
BRDY321974EA 65 EUR InStock
BRDY321974
Magicard™ Monochrome Black Film + Overcoat for Magicard™ 300
Farbbänder
Magicard™ 300 black monochrome ribbon with overcoat.
  • Create extra durable cards
  • Includes a cleaning roller for dust-free printing
Black mono colour dye film with overcoat protection.
Lieferumfang: Includes 1 dye film and 1 cleaning roller.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge