Absorbent socs, Allwik Universal
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
813789 813790
BRDY813789EA 110.6 EUR InStock
BRDY813789 BRDY813790
Absorbent socs, Allwik Universal
Absorptionsmaterial
These Allwik-Socs are used for absorbing leaks and spills around machines.
  • Absorbs a variety of industrial liquids including aggressive chemicals
  • Flexible design allows the soc to fit around any machine, corner or curve
  • Filled with universal meltblown polypropylene
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge