Labels, pipe marking
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
N002370 N004112 N004319 N004483 N004511 N004522 N002355 N000485 N003331 N003252 N003298 N005130 N005108 N005163 N004915
BRDYN002370EA 16.12 EUR InStock
BRDYN002370 BRDYN004112 BRDYN004319 BRDYN004483 BRDYN004511 BRDYN004522 BRDYN002355 BRDYN000485 BRDYN003331 BRDYN003252 BRDYN003298 BRDYN005130 BRDYN005108 BRDYN005163 BRDYN004915
Labels, pipe marking
Etiketten
These pipe marking labels are made of high quality polyester and meet with GHS (Global Harmonised System) regulations for chemical substances and mixtures.
  • Promotes safety and simplifies maintenance
  • Arrows are pre-cut, and hence does not requires scissors additionally
  • Resistant to temperatures from –40 to +120 °C
  • Resistant to ultraviolet rays and conditions
The markers have a frame that contrasts with the color of the text to increase visibility.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge