SPC® Oil-only Absorbents
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SKO-1100
BRDYSKO-1100EA 0 EUR InStock
BRDYSKO-1100
SPC® Oil-only Absorbents
Absorptionsmaterial
Drop front spill kit designed for larger spills of oil and petroleum based liquids. Top and drop front lid provides easy access to the absorbents inside the kit.
  • Absorbs up to 972 litres (full content of an IBC)
  • Absorbents and spill control
  • Drop front for fast and easy access
Wheeled to easily manoeuvre to location of the spill.
Lieferumfang: Includes: 300 ENV200-M pads, 2 OP15-E rolls, 4 ENV510 booms, 30 OIL430 socs, 10 OIL 124 socs, 20 OIL1818 pillows, 1 pair SPK-HANDSCH gloves, 1 pair SPK-BRIL goggles, 50 disposal bags.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge