Magnetrührstäbe, PTFE, oktagonal, mit Mittelring
Lieferant: Brand
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
137607 137608 137610 137612 137615 137617 137618 137620 137622 137625
BRND137607EA 47.5 EUR InStock
BRND137607 BRND137608 BRND137610 BRND137612 BRND137615 BRND137617 BRND137618 BRND137620 BRND137622 BRND137625
Magnetrührstäbe, PTFE, oktagonal, mit Mittelring
Rührer Magnetrührstäbchen
These octagonal shaped PTFE magnetic stirrer bars are ideal for vessels with slightly uneven bottoms. The angular surface creates high turbulence even at low speeds.
Designed with pivot ring structure which reduces the contact surface and enables the stirrer to adopt the optimum position.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge