Mixing measuring cylinders, tall form, class A, borosilicate glass, BLAUBRAND®
Lieferant: Brand
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
232408 232428 232420 232438 232448
BRND232408EA 53.7 EUR InStock
BRND232408 BRND232428 BRND232420 BRND232438 BRND232448
Mixing measuring cylinders, tall form, class A, borosilicate glass, BLAUBRAND®
Zylinder Messzylinder
Borosilicate glass 3.3, class A, conformity certified.
  • Calibrated to contain (TC, In)
  • Hexagonal glass base, PP stopper (except for 612-5028 and 612-5029 octagonal PE stopper)
  • Ring marks at major graduations, marks and inscriptions in high contrast blue enamel
* Height without stopper
Zertifizierungen: DIN EN ISO 4788
Bestellinformation: Batch certificate included.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge