Captive Spray 2 Emitter
Lieferant: BRUKER DALTRONIK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1811112
BRUK1811112
Captive Spray 2 Emitter
Ionisatoren
Our ready-to-use fused silica emitter offers a straightforward plug-and-play setup, ensuring hassle-free connection. Available in two internal diameters, our emitters cover both, high-throughput applications and the specific needs of plasma or single-cell proteomics.
  • Already attached ZDV union for easier handling
  • For use with the captivespray source, captivespray 2 source or captivespray ultra source
The 10 µm diameter is recommended for applications in the low nano flow range (50 to 450 nl/min).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge