UHPLC Columns, PepSep Series
Lieferant: BRUKER DALTRONIK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1893470
BRUK1893470EA 1270 EUR InStock
BRUK1893470
UHPLC Columns, PepSep Series
Chromatographiesäulen
Experience the true potential of your proteomics applications with our high-performance liquid chromatography C18 reversed phase columns. PepSep columns are created with industry leading materials and connections to accommodate all your chromatographic separation needs.
  • Patented female ultra-high pressure (UHP) connector for smooth, safe, and easy connections to the high-pressure end of the column
  • UHP connection is tested to be used up to 1000 bar
  • patented nanoConnect (nC) connector on the low-pressure end of the column makes union and emitter connections quick, reliable, and straightforward
  • nC connector ensures a zero dead volume connection
Discover the perfect solution for your Proteomics applications within our diverse PepSep column portfolio. With a wide range of columns available, featuring different lengths, inner diameters, and particle sizes, we are determined to meet your unique needs. Discover the perfect column for your specific requirements and revolutionize your proteomics analysis today.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge