MALDI AnchorChip™ Targets
Lieferant: BRUKER DALTRONIK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
8280790
BRUK8280790EA 3640 EUR InStock
BRUK8280790
MALDI AnchorChip™ Targets
Mikrotiterplatten Immunologie-Mikrotiterplatten
Patented AnchorChip™ targets are polished steel targets with hydrophilic sample and calibrant spots surrounded by a hydrophobic barrier for MALDI sample preparation.
  • Spot Ø: 800 µm
  • Sample spots: 384
  • Calibrant spots: 96
  • Autoteaching
Sample positions on contain 'anchors', hydrophilic patches surrounded by a hydrophobic ring. The 'anchor' localises droplets at the sample position and the hydrophobic ring prevents sample spreading and concentrates the sample into a spot between 0,4 and 2 mm in diameter. The localisation effect ensures that every single laser shot fired throughout an automatic run will hit a sample spot. This significantly increases the efficiency of the MALDI acquisition process.
Compatible with autoflex, ultraflex , rapifleX, timsTOF fleX, scimaX and solariX series instruments.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge