Optimised Filling Tools for MAS Rotors
Lieferant: BRUKER DALTRONIK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
H153429 H153427 H153428 H153878 H156143
BRUKH153429EA 0 EUR InStock
BRUKH153429 BRUKH153427 BRUKH153428 BRUKH153878 BRUKH156143
Optimised Filling Tools for MAS Rotors
Werkzeugsätze
For 0,7 mm rotors, Bruker offers specially designed workflow optimised toolsets to support handling of these tiny pars alongside a special preparation video describing the whole procedure.
The toolset for the small diameter is also colour coded to make it easier to identify the tools for the top and bottom caps.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge