MagSorb-16 Magnetic Rack for Manual Nucleic Acid Extraction
Lieferant: BIOSAN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BS-010601
BSANBS-010601EA 411 EUR InStock
BSANBS-010601
MagSorb-16 Magnetic Rack for Manual Nucleic Acid Extraction
Gestelle Trenngestelle, magnetisch
Biosan presents the complete line of necessary instrumentation to utilise magnetic bead extraction kits and protocols from various manufacturersand meet the most demanding user requirements.
The one of the foundations of this line is MagSorb-16 which is a magnetic rack that easily accommodates up to 16 single usetubes (1,5 to 2 ml). Therack consists of following parts: tube mounting racksand magnetic stand. Magnetic beads based NA extraction methods offer quick and efficient separation of genetic material from cellular leftovers.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge