Anti-SLC7A3 Rabbit Polyclonal Antibody
Lieferant: BosterBio
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
A09720-1
BSBTA09720-1EA 552 EUR InStock
BSBTA09720-1
Anti-SLC7A3 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Cationic amino acid transporter 3(SLC7A3) detection. Tested with WB in Human.
Type: Primary
Antigen: SLC7A3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge