Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-S-100 (beta-subunit) Mouse Monoclonal Antibody [clone: SA-12]

Lieferant: BosterBio

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

MA1088
BSBTMA1088EA 552 EUR InStock
BSBTMA1088
Anti-S-100 (beta-subunit) Mouse Monoclonal Antibody [clone: SA-12]
Antikörper
Mouse IgG monoclonal antibody for S-100 (beta-subunit), S100 calcium binding protein B (S100B) detection. Tested with IHC-P in Human;mouse;rat. No cross reactivity with other proteins.

S100 protein is a 21,000-Da component first isolated from brain by Moore(1965). Immunocytochemical studies demonstrated that S100 protein is produced by a wide variety of normal and neoplastic cells of mesodermal, neuroectodermal, and epithelial origin(Herrera et al., 1988). The S100 molecule is a dimer; thus there are 3 forms of S100: alpha-alpha, known as S-100a(0); alpha-beta, known as S-100a; and beta-beta, known as S-100b. S100 genes are clustered on human chromosome 1q21. S-100 protein expression by primary and metastatic adenocarcinomas.

1.2% sodium acetate or neutral PBS. If 1ml of PBS is used, the antibody concentration will be 100ug/ml.

Mouse ascites fluid, 1.2% sodium acetate, 2mg BSA, with 0.01mg NaN3 as preservative.

Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: S-100 (beta-subunit)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rabbit, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge