Rabbit IgG polyclonal antibody for Aldehyde dehydrogenase, mitochondrial(ALDH2) detection. Tested with WB, IHC-P in Human Rat. 1 * 0,1 mg
Lieferant: BOSTER BIOLOGICAL TECHNOLOGY
Anti-ALDH2 Rabbit Polyclonal Antibody
PRODUKT
Erweitert / Kompakt
Beschreibung Katalognummer Verpackungseinheit Menge
Rabbit IgG polyclonal antibody for Aldehyde dehydrogenase, mitochondrial(ALDH2) detection. Tested with WB, IHC-P in Human Rat. 1 * 0,1 mg BSBTPA2073 1 * 0,1 mg
Hersteller-Artikelnummer:: PA2073
Lokale Artikelnummer:: BSBTPA2073
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder