Anti-DDAH2 Rabbit Polyclonal Antibody
Lieferant: BosterBio
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PB10001
BSBTPB10001EA 552 EUR InStock
BSBTPB10001
Anti-DDAH2 Rabbit Polyclonal Antibody
Antikörper
DDAH2 is known as dimethylarginine dimethylaminohydrolase 2 which is mapped to 6p21.3 by radiation hybrid and FISH analysis. This gene encodes a dimethylarginine dimethylaminohydrolase. DDAH2 functions in nitric oxide generation by regulating the cellular concentrations of methylarginines, which in turn inhibit nitric oxide synthase activity. The protein may be localized to the mitochondria. Alternative splicing resulting in multiple transcript variants.
1. Leiper, J. M,, Santa Maria, J., Chubb, A., MacAllister, R. J., Charles, I. G., Whitley, G. S., Vallance, P. Identification of two human dimethylarginine dimethylaminohydrolases with distinct tissue distributions and homology with microbial arginine deiminases.Biochem. J. 343: 209-214, 1999.
2. Smith, C. L., Birdsey, G. M., Anthony, S., Arrigoni, F. I., Leiper, J. M., Vallance, P. Dimethylarginine dimethylaminohydrolase activity modulates ADMA levels, VEGF expression, and cell phenotype. Biochem. Biophys. Res. Commun. 308: 984-989, 2003.
3. Tran, C. T. L., Fox, M. F., Vallance, P., Leiper, J. M. Chromosomal localization, gene structure, and expression pattern of DDAH1: comparison with DDAH2 and implications for evolutionary origins. Genomics 68: 101-105, 2000.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge