Anti-ITLN1 Rabbit Polyclonal Antibody
Lieferant: BosterBio
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PB10004
BSBTPB10004EA 552 EUR InStock
BSBTPB10004
Anti-ITLN1 Rabbit Polyclonal Antibody
Antikörper
Intelectin-1, also known as omentin, is an intelectin encoded in humans by the ITLN1 gene. This gene is mapped to chromosome 1q21.3-q22 by genomic sequence analysis. It is expressed on multiple cell types and appears to participate in insulin signaling and microbe recognition. Intelectin-1 functions both as a receptor for bacterial arabinogalactans and for lactoferrin. Having conserved ligand binding site residues, both human and mouse intelectin-1 bind the exocyclic vicinal diol of carbohydrate ligands such as galactofuranose.
1. "Entrez Gene: ITLN1 intelectin 1 (galactofuranose binding)".
2. Lee JK, Schnee J, Pang M, Wolfert M, Baum LG, Moremen KW, Pierce M (Feb 2001). "Human homologs of the Xenopus oocyte cortical granule lectin XL35". Glycobiology 11 (1): 65–73.
3. Tsuji S, Uehori J, Matsumoto M, Suzuki Y, Matsuhisa A, Toyoshima K, Seya T (Jun 2001). "Human intelectin is a novel soluble lectin that recognizes galactofuranose in carbohydrate chains of bacterial cell wall". J Biol Chem 276 (26): 23456–63.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge