Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Perforin Polyclonal Antibody

Lieferant: BosterBio

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

PB9155
BSBTPB9155EA 552 EUR InStock
BSBTPB9155
Anti-Perforin Polyclonal Antibody
Antikörper
Polyclonal antibody for PERFORIN/PRF1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. PERFORIN/PRF1 information: Molecular Weight: 61377 MW; Subcellular Localization: Cytoplasmic granule lumen. Secreted. Cell membrane; Multi-pass membrane protein. Endosome lumen. Stored in cytoplasmic granules of cytolytic T-lymphocytes and secreted into the cleft between T-lymphocyte and target cell. Inserts into the cell membrane of target cells and forms pores. Membrane insertion and pore formation requires a major conformation change. May be taken up via endocytosis involving clathrin-coated vesicles and accumulate in a first time in large early endosomes.

Type: Primary
Antigen: Perforin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge