Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Kv4.3 Polyclonal Antibody

Lieferant: BosterBio

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

PB9229
BSBTPB9229EA 552 EUR InStock
BSBTPB9229
Anti-Kv4.3 Polyclonal Antibody
Antikörper
Polyclonal antibody for KV4.3/KCND3 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. KV4.3/KCND3 information: Molecular Weight: 73451 MW; Subcellular Localization: Membrane; Multi-pass membrane protein. Cell membrane, sarcolemma . Cell projection, dendrite ; Tissue Specificity: Highly expressed in heart and brain, in particular in cortex, cerebellum, amygdala and caudate nucleus. Detected at lower levels in liver, skeletal muscle, kidney and pancreas. Isoform 1 predominates in most tissues. Isoform 1 and isoform 2 are detected at similar levels in brain, skeletal muscle and pancreas.

Type: Primary
Antigen: Kv4.3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge