Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Fetuin A Polyclonal Antibody

Lieferant: BosterBio

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

PB9242
BSBTPB9242EA 552 EUR InStock
BSBTPB9242
Anti-Fetuin A Polyclonal Antibody
Antikörper
Polyclonal antibody for 2017 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Mouse. 2017 information: Molecular Weight: 37326 MW; Subcellular Localization: Secreted; Tissue Specificity: Liver is the major site of synthesis, but fetuin is also expressed in limb buds and other extrahepatic tissues during development.

Type: Primary
Antigen: Fetuin A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge