Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-IL-15 Polyclonal Antibody

Lieferant: BosterBio

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

PB9244
BSBTPB9244EA 552 EUR InStock
BSBTPB9244
Anti-IL-15 Polyclonal Antibody
Antikörper
Polyclonal antibody for IL-15/IL15 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: ELISA. Reactive species: Human. IL-15/IL15 information: Molecular Weight: 18086 MW; Subcellular Localization: Isoform IL15-S48AA: Secreted; Tissue Specificity: Most abundant in placenta and skeletal muscle. It is also detected in the heart, lung, liver and kidney. IL15- S21AA is preferentially expressed in tissues such as testis and thymus.

Type: Primary
Antigen: IL-15
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge