Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SFRP2 Rabbit Polyclonal Antibody

Lieferant: BosterBio

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

PB9362
BSBTPB9362EA 552 EUR InStock
BSBTPB9362
Anti-SFRP2 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Secreted frizzled-related protein 2(SFRP2) detection. Tested with WB, ELISA in Human.

SFRP2 is also known as FRP-2, SARP1 or SDF-5. It is Expressed in adipose tissue, heart, brain, skeletal muscle, pancreas, thymus, prostate, testis, ovary, small intestine and colon. Highest levels in adipose tissue, small intestine and colon. This gene encodes a member of the SFRP family that contains a cysteine-rich domain homologous to the putative Wnt-binding site of Frizzled proteins. SFRPs act as soluble modulators of Wnt signaling. Methylation of this gene is a potential marker for the presence of colorectal cancer. 

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.

WB: The detection limit for SFRP2 is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: SELP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: Yes
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge