Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ATXN3 Rabbit Polyclonal Antibody

Lieferant: BosterBio

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

PB9423
BSBTPB9423EA 552 EUR InStock
BSBTPB9423
Anti-ATXN3 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Ataxin-3(ATXN3) detection. Tested with WB, IHC-P in Human;Rat.

ATXN3 (Ataxin 3), also known as AT3, MJD GENE, MJD1, SCA3 GENE, ATX3, JOS, Spinocerebellar ataxia-3, Machado-Joseph disease protein 1, is a protein that in humans is encoded by the ATXN3 gene. ATXN3 ranges in size from 360 to 374 amino acids. Using Northern blot analysis showed that ATXN3 mRNA was ubiquitously expressed in human tissues. They detected at least 4 ATXN3 transcripts of 1.4, 1.8, 4.5, and 7.5 kb and suggested that the different mRNA species probably result from differential splicing and polyadenylation. Machado-Joseph disease, also known as spinocerebellar ataxia-3, is an autosomal dominant neurologic disorder. The protein encoded by the ATXN3 gene contains (CAG)n repeats in the coding region, and the expansion of these repeats from the normal 13-36 to 68-79 is the cause of Machado-Joseph disease. There is an inverse correlation between the age of onset and CAG repeat numbers. Alternatively spliced transcript variants encoding different isoforms have been described for this gene. Ataxin-3 interacted with 2 human homologs of the yeast DNA repair protein RAD23, HHR23A (RAD23A) and HHR23B (RAD23B). Both normal and mutant ataxin-3 proteins interacted with the ubiquitin-like domain at the N terminus of the HHR23 proteins, which is a motif important for nucleotide excision repair. However, in HEK 293 cells, HHR23A was recruited to intranuclear inclusions formed by the mutant ataxin-3 through its interaction with ataxin-3.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.

Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: CCNA2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge