Anti-GJC1 Rabbit Polyclonal Antibody
Lieferant: BosterBio
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PB9624
BSBTPB9624EA 552 EUR InStock
BSBTPB9624
Anti-GJC1 Rabbit Polyclonal Antibody
Antikörper
Rabbit IgG polyclonal antibody for Gap junction gamma-1 protein(GJC1) detection. Tested with WB in Human;Rat.
Gap junction gamma-1 protein (GJC1), also known as gap junction alpha-7 protein (GJA7) or connexin 45 (Cx45), is a protein that in humans is encoded by the GJC1 gene. The International Radiation Hybrid Mapping Consortium mapped the GJA7 gene to chromosome 17q21.31. This gene is a member of the connexin gene family. The encoded protein is a component of gap junctions, which are composed of arrays of intercellular channels that provide a route for the diffusion of low molecular weight materials from cell to cell.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge