Anti-AGO4 Rabbit Polyclonal Antibody
Lieferant: BosterBio
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PB9979
BSBTPB9979EA 552 EUR InStock
BSBTPB9979
Anti-AGO4 Rabbit Polyclonal Antibody
Antikörper
AGO4 (Argonaute 4) is also known as EIF2C4. This gene encodes a member of the Argonaute family of proteins which play a role in RNA interference. The encoded protein is highly basic containing PAZ and PIWI domains, and it may play a role in short-interfering-RNA-mediated gene silencing. This gene is located on chromosome 1 in a cluster of closely related family members including argonaute 3, and eukaryotic translation initiation factor 2C, 1.
1. Carmell, M. A., Xuan, Z., Zhang, M. Q., Hannon, G. J. The argonaute family: tentacles that reach into RNAi, developmental control, stem cell maintenance, and tumorigenesis. Genes Dev. 16: 2733-2742, 2002.
2. Nagase, T., Kikuno, R., Nakayama, M., Hirosawa, M., Ohara, O. Prediction of the coding sequences of unidentified human genes. XVIII. The complete sequences of 100 new cDNA clones from brain which code for large proteins in vitro. DNA Res. 7: 273-281, 2000.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge