Anti-CA2 Rabbit Polyclonal Antibody
Lieferant: BosterBio
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PB9989
BSBTPB9989EA 552 EUR InStock
BSBTPB9989
Anti-CA2 Rabbit Polyclonal Antibody
Antikörper
CA2 is a cytosolic enzyme with the highest activity among all known CAs. The carbonic anhydrases (ACs) form a family of enzymes that catalyze the rapid interconversion of carbon dioxide and water to bicarbonate and protons (or vice versa), a reversible reaction that occurs relatively slowly in the absence of a catalyst. Mutations in the CA2 gene result in the CA II deficiency syndrome, an autosomal recessive disorder that produces osteopetrosis, renal tubular acidosis and cerebral calcification. This gene is mapped to 8q22.
1. Aramaki, S., Yoshida, I., Yoshino, M., Kondo, M., Sato, Y., Noda, K., Jo, R., Okue, A., Sai, N., Yamashita, F. Carbonic anhydrase II deficiency in three unrelated Japanese patients. J. Inherit. Metab. Dis. 16: 982-990, 1993.
2. Laitala, T., Vaananen, H. K. Inhibition of bone resorption in vitro by antisense RNA and DNA molecules targeted against carbonic anhydrase II or two subunits of vacuolar H(+)-ATPase. J. Clin. Invest. 93: 2311-2318, 1994.
3. Venta, P. J., Welty, R. J., Johnson, T. M., Sly, W. S., Tashian, R. E. Carbonic anhydrase II deficiency syndrome in a Belgian family is caused by a point mutation at an invariant histidine residue (107his-to-tyr): complete structure of the normal human CA II gene. Am. J. Hum. Genet. 49: 1082-1090, 1991.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge