Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CK1 Chicken Polyclonal Antibody

Lieferant: Biosensis

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

C-1371-100
BSENC-1371-100EA 312 EUR InStock
BSENC-1371-100
Anti-CK1 Chicken Polyclonal Antibody
Antikörper
Casein kinases are operationally defined by their preferential utilization of acidic proteins such as caseins as substrates. It can phosphorylate a large number of proteins. Participates in Wnt signaling. Phosphorylates CTNNB1 at 'Ser-45'. May phosphorylate PER1 and PER2. May play a role in segregating chromosomes during mitosis. May play a role in keratin cytoskeleton disassembly and thereby, it may regulate epithelial cell migration. (Reference: uniprot.org)

Western Blotting (WB): 1:5,000 - 1:10,000. Casein kinase 1 alpha has a predicted molecular weight of 38 kDa.

Immunocytochemistry (ICC): 1:500-1:1,000.

Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Shortest isoform of recombinant full length CK1a.
Host: Chicken
Isotype:
Reactivity: Human;Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge