Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Neurofilament Light Chicken Polyclonal Antibody

Lieferant: Biosensis

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

C-1390-50
BSENC-1390-50EA 312 EUR InStock
BSENC-1390-50
Anti-Neurofilament Light Chicken Polyclonal Antibody
Antikörper
Neurofilaments are composed of three intermediate filament proteins: light (~68 kDa), medium (~160 kDa) and heavy (~200 kDa), which are involved in the maintenance of the neuronal caliber. Neurofilament light (NF68 or NF-L) is the most abundant of the three proteins.

The specificity of this antibody has been confirmed by IC and western blot. The antibody reacts excellently in western blots against extracts of rat spinal cord, brain stem, and other brain regions.
Application Information:
Western Blotting (WB) and Immunocytochemistry (IC). A dilution of 1:5,000 - 1:10,000 is recommended for WB. A dilution of 1:1,000 - 1:5,000 is recommended for IC. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Neurofilament Light
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Chicken
Isotype: IgY
Reactivity: Human, Rat, Mouse, Cat, Chicken

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge