Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-C-reactive Protein Chicken Polyclonal Antibody

Lieferant: Biosensis

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

C-1509-100
BSENC-1509-100EA 321 EUR InStock
BSENC-1509-100
Anti-C-reactive Protein Chicken Polyclonal Antibody
Antikörper
C-reactive protein has several roles associated with host defence such as; promoting agglutination, bacterial capsular swelling, phagocytosis and complement fixation through its calcium-dependent binding to phosphorylcholine. It can interact with DNA and histones and may scavenge nuclear material released from damaged circulating cells. COFACTOR: Binds 2 calcium ions per subunit. C-reactive protein exists as a homopentamer. There are 2 alternatively spliced isoforms. C-reactive protein is found in plasma and its concentration increases greatly during acute phase response to tissue injury, infection or other inflammatory stimuli. It is induced by IL-1 and IL-6.

The antibody reacts specifically with human C-reactive protein (CRP) and likely reacts with C-reactive protein from mouse, rat, pig, and other mammalian species. CRP monomer runs as 21-25kDa in fully reduced westerns. CRP in native state is a homopentameric protein of 115-118 kDa in size non-reduced. Human serum that has been separated from whole blood and concentrated though a 3 kDa centrifugal concentrator can serve as a positive control. The antibody also detects human C-reactive protein in ELISA.
Application Information:
ELISA and WB. Suggested dilution of 1:2,000-1:5,000. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: C-reactive Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Chicken
Isotype: IgY
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge