Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Acetyl Lysine (AcK) Chicken Polyclonal Antibody

Lieferant: Biosensis

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

C-1516-100
BSENC-1516-100EA 379 EUR InStock
BSENC-1516-100
Anti-Acetyl Lysine (AcK) Chicken Polyclonal Antibody
Antikörper
Lysine acetylation of histones and non-histone proteins plays an important part in many cellular processes such as chromatin and nuclear signaling, transcription, gene silencing, cell cycle progression, apoptosis, differentiation, DNA replication and repair.

The antibody specifically binds to proteins with acetylated lysine residues. The antibody has been testeed in western blot and ELISA and is expected to also work in immunoprecipitation if a rabbit-anti-chicken IgG or other IgY binding capture unit is used, however IP is currently untested for this antibody.
Application Information:
ELISA and WB. Suggested dilution of 1:1,000-1:5,000. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: Acetyl Lysine (AcK)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Chicken
Isotype: IgY
Reactivity: Many Species

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge