Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-G-Protein subunit beta-3 Chicken Polyclonal Antibody

Lieferant: Biosensis

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

C-1520-100
BSENC-1520-100EA 321 EUR InStock
BSENC-1520-100
Anti-G-Protein subunit beta-3 Chicken Polyclonal Antibody
Antikörper
G-Protein beta3 (GNB3) is a guanine nucleotide-binding protein (G protein). G-proteins are involved as a modulator or transducer in various transmembrane signaling systems (Ref: SWISSPROT).

The antibody reacts specifically with GNB3 protein of human, mouse, and rat. Because of homology it is expects to react with other mammalian species as well. The antibody detects a prominent band ~38 kDa in western blot with extracts from human adipose tissue.
Application Information:
WB. Suggested dilution at 1:1,000 to 1:5,000. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: G-Protein subunit beta-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human GNB3 amino acids 216-230.
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge