Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Hormone Sensitive Lipase Chicken Polyclonal Antibody

Lieferant: Biosensis

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

C-1523-500
BSENC-1523-500EA 321 EUR InStock
BSENC-1523-500
Anti-Hormone Sensitive Lipase Chicken Polyclonal Antibody
Antikörper
Hormone Sensitive Lipase (HSL) hydrolyzes stored triglycerides to free fatty acids in adipose tissue and heart. In steroidogenic tissues, HSL principally converts cholesteryl esters to free cholesterol for steroid hormone production (ref: SWISSPROT).

This antibody detects HSL at approx 83-84 kDa in various fat cell lysates from mouse and rat. Additional weaker band may appear at approx 40 kDa (unknown).
Application Information:
WB. Suggested dilution at 1:1,000 to 1:5,000. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: Hormone Sensitive Lipase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple peptide epitopes cooresponding to amino acids 617-828, 533-544 rat, and 191-202 of Mouse HSL protein.
Host: Chicken
Isotype: IgY
Reactivity: Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge