Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Fatty acid-binding Protein Chicken Polyclonal Antibody

Lieferant: Biosensis

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

C-1527-100
BSENC-1527-100EA 321 EUR InStock
BSENC-1527-100
Anti-Fatty acid-binding Protein Chicken Polyclonal Antibody
Antikörper
Fatty acid-binding protein, adipocyte (ALBP) is a lipid transport protein which binds long chain fatty acids and other hydrophobic ligands and delivers them to their receptors in the nucleus. ALBP is found in the cytoplasm and nucleus of adipocytes.

This antibody detects human Fatty acid-binding protein, adipocyte at approx 16 kDa. The antibody may detect ALBP from other species because of protein homology
Application Information:
WB. Suggested dilution of 1:2,000-1:5,000. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: Fatty acid-binding Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Maps to amino acids 97-111 of Human FABP4 protein
Host: Chicken
Isotype: IgY
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge